Assay Details
Target Gene Details
Entrez Gene ID: | 129138 |
Gene Name: | ankyrin repeat domain 54 |
Gene Aliases: |
LIAR |
Location: |
Chr.22:37830854-37848994 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD54 | NM_138797.2 | NP_620152.1 | ||
| NR_036556.1 | ||||
| XM_006724136.1 | XP_006724199.1 | |||
| XM_006724137.1 | XP_006724200.1 | |||
| XM_011529877.2 | XP_011528179.1 | |||
| XM_011529878.2 | XP_011528180.1 | |||
| XM_011529879.2 | XP_011528181.1 | |||
| XM_011529880.2 | XP_011528182.1 | |||
| XM_011529882.2 | XP_011528184.1 | |||
| XM_011529883.2 | XP_011528185.1 | |||
| XM_011529884.2 | XP_011528186.1 | |||
| XM_017028590.1 | XP_016884079.1 | |||
| XM_017028591.1 | XP_016884080.1 | |||
| XM_017028593.1 | XP_016884082.1 | |||
| AK127583.1 | BAC87043.1 | |||
| BC014641.1 | AAH14641.1 | |||
| BC066909.1 | AAH66909.1 | |||
| BX401122.2 | ||||
| CR456471.1 | CAG30357.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758838 | Chr.22:37607870 - 37869871 on Build GRCh38 | Loss |
|
| esv3893482 | Chr.22:37832559 - 37927500 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map