Assay Details
Target Gene Details
Entrez Gene ID: | 4486 |
Gene Name: | macrophage stimulating 1 receptor |
Gene Aliases: |
CD136, CDw136, PTK8, RON |
Location: |
Chr.3:49886471-49903637 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MST1R | NM_001244937.2 | NP_001231866.1 | ||
| NM_001318913.1 | NP_001305842.1 | |||
| NM_002447.3 | NP_002438.2 | |||
| NR_134919.1 | ||||
| XM_005265170.4 | XP_005265227.2 | |||
| XM_011533739.2 | XP_011532041.1 | |||
| XM_011533740.2 | XP_011532042.1 | |||
| XM_011533741.2 | XP_011532043.1 | |||
| XM_011533742.2 | XP_011532044.1 | |||
| XM_011533743.2 | XP_011532045.1 | |||
| XM_011533744.2 | XP_011532046.1 | |||
| AK303234.1 | ||||
| BU542597.1 | ||||
| EU826584.1 | 10 | 2630 | ||
| HQ908652.1 | ||||
| JN689381.1 | ||||
| JN689382.1 | ||||
| JN983806.1 | ||||
| X70040.1 | CAA49634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834688 | Chr.3:49773682 - 49981866 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map