Assay Details
Target Gene Details
Entrez Gene ID: | 10623 |
Gene Name: | RNA polymerase III subunit C |
Gene Aliases: |
RPC3, RPC62 |
Location: |
Chr.1:145823183-145844402 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3C | NM_006468.7 | 1 | 176 | NP_006459.3 |
| XM_005272927.3 | 1 | 1062 | XP_005272984.1 | |
| BC002586.1 | AAH02586.1 | |||
| BC004424.1 | 1 | 69 | AAH04424.1 | |
| BI831011.1 | 1 | 176 | ||
| U93867.1 | AAB63675.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015025 | Chr.1:145642988 - 145851919 on Build GRCh38 | Gain |
|
| nsv831381 | Chr.1:145784568 - 145960930 on Build GRCh38 | Gain |
|
| esv3894089 | Chr.1:145672100 - 145824903 on Build GRCh38 | Loss |
|
| dgv531n54 | Chr.1:145689573 - 145824903 on Build GRCh38 | Loss |
|
| dgv33n27 | Chr.1:145687602 - 145824903 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map