Assay Details
Target Gene Details
Entrez Gene ID: | 5818 |
Gene Name: | nectin cell adhesion molecule 1 |
Gene Aliases: |
CD111, CLPED1, ED4, HIgR, HV1S, HVEC, OFC7, PRR, PRR1, PVRL1, PVRR, PVRR1, SK-12, nectin-1 |
Location: |
Chr.11:119638098-119728725 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECTIN1 | NM_203285.1 | NP_976030.1 | ||
| AF110314.1 | AAD04944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627893 | Chr.11:119634114 - 119639011 on Build GRCh38 | Loss |
|
| nsv509445 | Chr.11:119609244 - 119695172 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map