Assay Details
Target Gene Details
Entrez Gene ID: | 51552 |
Gene Name: | RAB14, member RAS oncogene family |
Gene Aliases: |
FBP, RAB-14 |
Location: |
Chr.9:121178137-121202087 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB14 | NM_016322.3 | 8 | 2448 | NP_057406.2 |
| AF052113.1 | 1 | 895 | ||
| AK000447.1 | 1 | 762 | ||
| AK023524.1 | 8 | 2230 | BAB14598.1 | |
| AK090889.1 | 8 | 2247 | ||
| AL110256.1 | 1 | 831 | ||
| AL162081.1 | 8 | 2175 | CAB82414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950937 | Chr.9:121178823 - 121200622 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map