Assay Details
Target Gene Details
Entrez Gene ID: | 115584 |
Gene Name: | solute carrier family 5 member 11 |
Gene Aliases: |
KST1, RKST1, SGLT6, SMIT2 |
Location: |
Chr.16:24832784-24911711 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A11 | NM_001258411.2 | 2 | 244 | NP_001245340.1 |
| NM_001258412.2 | 2 | 244 | NP_001245341.1 | |
| NM_001258413.2 | 2 | 244 | NP_001245342.1 | |
| NM_001258414.1 | 2 | 244 | NP_001245343.1 | |
| NM_052944.4 | 1 | 95 | NP_443176.2 | |
| XM_005255080.2 | 2 | 123 | XP_005255137.1 | |
| XM_006721015.2 | 2 | 367 | XP_006721078.1 | |
| XM_011545725.2 | 1 | 279 | XP_011544027.2 | |
| XM_017022900.1 | 2 | 439 | XP_016878389.1 | |
| XM_017022901.1 | 2 | 1149 | XP_016878390.1 | |
| XM_017022902.1 | 2 | 278 | XP_016878391.1 | |
| XM_017022903.1 | 2 | 182 | XP_016878392.1 | |
| XM_017022904.1 | 2 | 366 | XP_016878393.1 | |
| XM_017022905.1 | 2 | 439 | XP_016878394.1 | |
| XM_017022906.1 | 2 | 366 | XP_016878395.1 | |
| XM_017022907.1 | 2 | 123 | XP_016878396.1 | |
| XM_017022908.1 | 2 | 182 | XP_016878397.1 | |
| XM_017022909.1 | 2 | 123 | XP_016878398.1 | |
| AK125267.1 | BAC86105.1 | |||
| AK295427.1 | 2 | 113 | ||
| AK298373.1 | ||||
| AK303691.1 | 2 | 112 | ||
| AY044906.1 | 1 | 95 | AAK97784.1 | |
| BC049385.1 | 2 | 114 | ||
| BC057780.1 | 2 | 113 | AAH57780.1 | |
| DC332171.1 | 2 | 244 |
More Information
Set Membership: |
|

Genomic Map