Assay Details
Target Gene Details
Entrez Gene ID: | 80829 |
Gene Name: | ZFP91 zinc finger protein |
Gene Aliases: |
DMS-8, DSM-8, FKSG11, PZF, ZFP-91, ZNF757 |
Location: |
Chr.11:58579114-58621550 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFP91 | NM_001197051.1 | 11 | 4464 | NP_001183980.1 |
| NM_053023.4 | 11 | 4467 | NP_444251.1 | |
| AB056107.1 | 11 | 4413 | BAB63373.1 | |
| AK025502.1 | 1 | 1449 |
Target Gene Details
Entrez Gene ID: | 386607 |
Gene Name: | ZFP91-CNTF readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:58579114-58625733 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFP91-CNTF | NR_024091.1 | |||
| AB057443.1 | BAB63374.1 |
More Information
Set Membership: |
|

Genomic Map