Assay Details
Target Gene Details
Entrez Gene ID: | 116138 |
Gene Name: | kelch domain containing 3 |
Gene Aliases: |
PEAS, dJ20C7.3 |
Location: |
Chr.6:43014103-43021298 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHDC3 | NM_057161.3 | 11 | 1587 | NP_476502.1 |
| NR_040101.1 | 10 | 1471 | ||
| AB055925.1 | 11 | 1451 | BAB63257.1 | |
| AK097719.1 | 9 | 1313 | BAC05149.1 | |
| AK290384.1 | 11 | 1443 | ||
| AK302336.1 | 10 | 1226 | ||
| BC000295.2 | 11 | 1407 | AAH00295.1 | |
| BC001789.1 | 11 | 1422 | AAH01789.1 | |
| BC001793.2 | 11 | 1385 | AAH01793.1 | |
| BC007296.1 | 11 | 1437 | AAH07296.1 | |
| BC009460.1 | AAH09460.1 | |||
| BC021546.1 | 11 | 1424 | AAH21546.1 | |
| BC041793.1 | 10 | 1325 | AAH41793.2 | |
| BC045612.1 | 10 | 1325 | AAH45612.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462927 | Chr.6:42948182 - 43110718 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
| nsv462930 | Chr.6:43002769 - 43110718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map