Assay Details
Target Gene Details
Entrez Gene ID: | 571 |
Gene Name: | BTB domain and CNC homolog 1 |
Gene Aliases: |
BACH-1, BTBD24 |
Location: |
Chr.21:29298795-29361896 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACH1 | NM_001186.3 | 3 | 1103 | NP_001177.1 |
| NM_206866.2 | 3 | 1127 | NP_996749.1 | |
| NR_027655.2 | 3 | 1063 | ||
| AB002803.1 | 3 | 1002 | BAA24932.1 | |
| AF026199.1 | 3 | 1127 | AAB84100.1 | |
| AK291021.1 | 3 | 1060 | ||
| AK313791.1 | 3 | 976 | ||
| BC063307.1 | 3 | 976 | AAH63307.1 | |
| CR450303.1 | 2 | 884 | CAG29299.1 |
More Information
Set Membership: |
|

Genomic Map