Assay Details
Target Gene Details
Entrez Gene ID: | 81831 |
Gene Name: | neuropilin and tolloid like 2 |
Gene Aliases: |
BTCL2, NEOT2 |
Location: |
Chr.16:47081520-47144025 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NETO2 | NM_001201477.1 | 9 | 2798 | NP_001188406.1 |
| NM_018092.4 | 9 | 2819 | NP_060562.3 | |
| XM_006721289.1 | 8 | 2705 | XP_006721352.1 | |
| XM_006721291.1 | 8 | 2684 | XP_006721354.1 | |
| XM_017023740.1 | 9 | 2533 | XP_016879229.1 | |
| AK001292.1 | 1 | 1313 | BAA91604.1 | |
| AK027478.1 | 1 | 1390 | BAB55141.1 | |
| AK027630.1 | 2 | 1446 | BAB55247.1 | |
| AL834404.1 | 6 | 2185 | CAD39066.1 | |
| AY358718.1 | 9 | 2716 | AAQ89080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map