Assay Details
Target Gene Details
Entrez Gene ID: | 90427 |
Gene Name: | Bcl2 modifying factor |
Gene Aliases: |
- |
Location: |
Chr.15:40087890-40108884 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BMF | NM_001003940.1 | 5 | 3446 | NP_001003940.1 |
| NM_001003942.1 | 3 | 3148 | NP_001003942.1 | |
| NM_001003943.2 | 4 | 3295 | NP_001003943.1 | |
| NM_033503.3 | 4 | 3297 | NP_277038.1 | |
| AK024472.1 | 4 | 3307 | ||
| BC060783.1 | 5 | 3444 | AAH60783.1 | |
| BC070043.1 | 5 | 3444 | ||
| BQ064116.1 | 1 | 665 |
More Information
Additional Information:
For this assay, SNP(s) [rs148885586] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map