Assay Details
Target Gene Details
Entrez Gene ID: | 645513 |
Gene Name: | septin 7 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.4:119454783-119499592 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC645513 | NR_037630.1 | |||
| BM544557.1 |
Target Gene Details
Entrez Gene ID: | 8654 |
Gene Name: | phosphodiesterase 5A |
Gene Aliases: |
CGB-PDE, CN5A, PDE5 |
Location: |
Chr.4:119494395-119629080 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE5A | NM_001083.3 | 21 | 4278 | NP_001074.2 |
| NM_033430.2 | 21 | 4109 | NP_236914.2 | |
| NM_033437.3 | 21 | 4061 | NP_246273.2 | |
| BC126233.1 | 21 | 4212 |
More Information
Set Membership: |
|

Genomic Map