Assay Details
Target Gene Details
Entrez Gene ID: | 4882 |
Gene Name: | natriuretic peptide receptor 2 |
Gene Aliases: |
AMDM, ANPRB, ANPb, ECDM, GUC2B, GUCY2B, NPRB, NPRBi, SNSK |
Location: |
Chr.9:35752945-35809731 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPR2 | NM_003995.3 | 19 | 2804 | NP_003986.2 |
| XM_005251478.3 | 19 | 3600 | XP_005251535.1 | |
| XM_005251479.4 | 21 | 6721 | XP_005251536.1 | |
| XM_011517890.2 | 20 | 7305 | XP_011516192.1 | |
| XM_011517891.2 | 21 | 7214 | XP_011516193.1 | |
| XM_011517892.2 | 22 | 7386 | XP_011516194.1 | |
| XM_011517893.2 | 21 | 7183 | XP_011516195.1 | |
| XM_011517895.2 | 14 | 1555 | XP_011516197.1 | |
| XM_017014745.1 | 21 | 6717 | XP_016870234.1 | |
| XM_017014746.1 | 21 | 6708 | XP_016870235.1 | |
| XM_017014747.1 | 20 | 6162 | XP_016870236.1 | |
| XM_017014748.1 | 21 | 7514 | XP_016870237.1 | |
| XM_017014749.1 | 21 | 6674 | XP_016870238.1 | |
| XM_017014750.1 | 20 | 6498 | XP_016870239.1 | |
| AJ005282.1 | 19 | 2804 | CAA06466.1 | |
| BC023017.1 | 14 | 1517 | AAH23017.1 | |
| BQ446138.1 | 4 | 644 | ||
| L13436.1 | 19 | 3454 |
Target Gene Details
Entrez Gene ID: | 26206 |
Gene Name: | sperm associated antigen 8 |
Gene Aliases: |
BS-84, CILD28, CT142, HSD-1, SMP1, SPAG3, hSMP-1 |
Location: |
Chr.9:35807785-35812262 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAG8 | NM_172312.1 | NP_758516.1 | ||
| BC019247.1 | AAH19247.1 | |||
| S83157.1 | AAB46833.1 | |||
| U12978.2 | AAA21129.2 |
More Information
Set Membership: |
|

Genomic Map