Assay Details
Target Gene Details
Entrez Gene ID: | 79974 |
Gene Name: | cadherin like and PC-esterase domain containing 1 |
Gene Aliases: |
C7orf58 |
Location: |
Chr.7:120988697-121297444 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPED1 | NM_001105533.1 | 1 | 303 | NP_001099003.1 |
| NM_024913.4 | 2 | 519 | NP_079189.4 | |
| XM_017012649.1 | 1 | 995 | XP_016868138.1 | |
| AK055624.1 | 1 | 643 | BAB70974.1 | |
| AK289978.1 | 2 | 516 | ||
| AL832619.2 | 2 | 474 | CAD89961.1 | |
| AL832638.2 | 2 | 343 | CAD89953.1 | |
| AY358220.1 | 1 | 95 | AAQ88587.1 | |
| AY780785.1 | 1 | 72 | AAV98355.1 | |
| BC046360.1 | 1 | 519 | AAH46360.1 | |
| BQ082412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831114 | Chr.7:120955570 - 121164456 on Build GRCh38 | Loss |
|

Genomic Map