Assay Details
Target Gene Details
Entrez Gene ID: | 100294362 |
Gene Name: | uncharacterized LOC100294362 |
Gene Aliases: |
- |
Location: |
Chr.17:80351830-80415168 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100294362 | NR_029376.1 | 2 | 961 | |
| AK125341.1 | 2 | 912 | BAC86138.1 |
Target Gene Details
Entrez Gene ID: | 57674 |
Gene Name: | ring finger protein 213 |
Gene Aliases: |
ALO17, C17orf27, KIAA1618, MYMY2, MYSTR, NET57 |
Location: |
Chr.17:80260820-80398786 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 39 - Intron 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF213 | NM_001256071.2 | NP_001243000.2 | ||
| XM_005257545.4 | XP_005257602.2 | |||
| XM_005257546.4 | XP_005257603.2 | |||
| XM_006721995.3 | XP_006722058.1 | |||
| XM_011525084.2 | XP_011523386.1 | |||
| XM_011525086.2 | XP_011523388.1 | |||
| XM_011525087.2 | XP_011523389.1 | |||
| XM_017024905.1 | XP_016880394.1 | |||
| AB537889.1 | ||||
| AK127358.1 | ||||
| AK127914.1 | BAC87188.1 | |||
| BX640932.1 | CAE45967.1 | |||
| BX647946.1 | CAH56189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3641372 | Chr.17:80351964 - 80357069 on Build GRCh38 | Loss |
|
| nsv952384 | Chr.17:80344901 - 80356400 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map