Assay Details
Target Gene Details
Entrez Gene ID: | 2166 |
Gene Name: | fatty acid amide hydrolase |
Gene Aliases: |
FAAH-1, PSAB |
Location: |
Chr.1:46394267-46413848 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAAH | NM_001441.2 | 1 | 82 | NP_001432.2 |
| BC093632.1 | AAH93632.1 | |||
| BC110404.1 | ||||
| BC111941.1 | AAI11942.1 | |||
| BG718115.1 | 1 | 89 | ||
| U82535.1 | AAB58505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829748 | Chr.1:46258836 - 46410450 on Build GRCh38 | Loss |
|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
| nsv546162 | Chr.1:46341925 - 46405089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map