Assay Details
Target Gene Details
Entrez Gene ID: | 84515 |
Gene Name: | minichromosome maintenance 8 homologous recombination repair factor |
Gene Aliases: |
C20orf154, POF10, dJ967N21.5 |
Location: |
Chr.20:5950652-6000941 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM8 | NM_001281520.1 | 1 | 113 | NP_001268449.1 |
| NM_001281521.1 | 1 | 113 | NP_001268450.1 | |
| NM_001281522.1 | 1 | 113 | NP_001268451.1 | |
| NM_032485.5 | 1 | 113 | NP_115874.3 | |
| NM_182802.2 | 1 | 113 | NP_877954.1 | |
| XM_011529387.2 | 1 | 76 | XP_011527689.1 | |
| XM_017028105.1 | 1 | 88 | XP_016883594.1 | |
| XM_017028106.1 | 1 | 75 | XP_016883595.1 | |
| AJ439063.1 | 1 | 113 | CAD27750.1 | |
| AK297806.1 |
More Information
Set Membership: |
|

Genomic Map