Assay Details
Target Gene Details
Entrez Gene ID: | 84623 |
Gene Name: | kin of IRRE like 3 (Drosophila) |
Gene Aliases: |
KIRRE, MRD4, NEPH2, PRO4502 |
Location: |
Chr.11:126423493-127003460 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIRREL3 | NM_001161707.1 | 9 | 1434 | NP_001155179.1 |
| NM_001301097.1 | 9 | 1434 | NP_001288026.1 | |
| NM_032531.3 | 9 | 1434 | NP_115920.1 | |
| XM_011543026.2 | 9 | 1178 | XP_011541328.1 | |
| XM_011543027.2 | 10 | 1639 | XP_011541329.1 | |
| XM_011543028.2 | 10 | 1498 | XP_011541330.1 | |
| XM_011543030.2 | 9 | 1175 | XP_011541332.1 | |
| XM_011543031.2 | 10 | 1633 | XP_011541333.1 | |
| XM_011543032.2 | 9 | 1175 | XP_011541334.1 | |
| XM_011543033.1 | 10 | 1639 | XP_011541335.1 | |
| XM_017018418.1 | 9 | 1729 | XP_016873907.1 | |
| XM_017018419.1 | 10 | 1639 | XP_016873908.1 | |
| XM_017018420.1 | 10 | 1498 | XP_016873909.1 | |
| AB058770.1 | 9 | 1323 | ||
| AF480410.1 | 9 | 1073 | AAN73042.1 | |
| AY358743.1 | 9 | 1374 | AAQ89103.1 | |
| BC101775.1 | 9 | 1159 | AAI01776.1 | |
| BC101801.1 | 9 | 1159 | AAI01802.1 |
Target Gene Details
Entrez Gene ID: | 6484 |
Gene Name: | ST3 beta-galactoside alpha-2,3-sialyltransferase 4 |
Gene Aliases: |
CGS23, NANTA3, SAT3, SIAT4, SIAT4C, ST3GalIV, STZ |
Location: |
Chr.11:126355645-126459601 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST3GAL4 | XM_011542959.1 | XP_011541261.1 | ||
| XM_011542960.2 | XP_011541262.1 |
More Information
Set Membership: |
|

Genomic Map