Assay Details
Target Gene Details
Entrez Gene ID: | 79589 |
Gene Name: | ring finger protein 128, E3 ubiquitin protein ligase |
Gene Aliases: |
GRAIL |
Location: |
Chr.X:106693838-106797016 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF128 | NM_024539.3 | NP_078815.3 | ||
| NM_194463.1 | 1 | 334 | NP_919445.1 | |
| AF394689.1 | 1 | 94 | AAK77554.1 | |
| AK074264.1 | 1 | 334 | BAB85033.1 | |
| AK126553.1 | BAC86589.1 | |||
| BC063404.1 | 1 | 173 | AAH63404.1 | |
| BI600046.1 | 1 | 294 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752312 | Chr.X:105705935 - 107123872 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map