Assay Details
Target Gene Details
Entrez Gene ID: | 5269 |
Gene Name: | serpin family B member 6 |
Gene Aliases: |
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3 |
Location: |
Chr.6:2948159-2972165 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB6 | NM_001195291.2 | 8 | 1577 | NP_001182220.2 |
| NM_001271822.1 | 7 | 1002 | NP_001258751.1 | |
| NM_001271823.1 | 7 | 1100 | NP_001258752.1 | |
| NM_001271824.1 | 7 | 1484 | NP_001258753.1 | |
| NM_001271825.1 | 7 | 1086 | NP_001258754.1 | |
| NM_001297699.1 | 7 | 1727 | NP_001284628.1 | |
| NM_001297700.1 | 7 | 1383 | NP_001284629.1 | |
| NM_004568.5 | 7 | 1442 | NP_004559.4 | |
| XM_011514672.1 | 9 | 1380 | XP_011512974.1 | |
| XM_011514673.1 | 7 | 1534 | XP_011512975.1 | |
| XM_011514674.2 | 7 | 1119 | XP_011512976.1 | |
| XM_011514676.2 | 7 | 929 | XP_011512978.1 | |
| XM_017010940.1 | 8 | 941 | XP_016866429.1 | |
| XM_017010941.1 | 6 | 2570 | XP_016866430.1 | |
| AB209563.1 | 6 | 1110 | BAD92800.1 | |
| AF116727.1 | 4 | 1663 | AAO15303.1 | |
| AK027149.1 | 6 | 2268 | ||
| AK057138.1 | 6 | 2801 | ||
| AK094983.1 | 8 | 1577 | ||
| AK098566.1 | 7 | 1099 | ||
| AK122895.1 | 5 | 1705 | ||
| AK122961.1 | 6 | 1718 | ||
| AK128637.1 | 5 | 3131 | ||
| AK314578.1 | 7 | 1161 | ||
| BC001394.2 | 7 | 1024 | AAH01394.1 | |
| BC098564.1 | 7 | 880 | AAH98564.1 | |
| BX538343.1 | 7 | 5154 | CAD98106.1 | |
| Z22658.1 | 7 | 873 | CAA80373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950760 | Chr.6:2946367 - 2957166 on Build GRCh38 | Deletion |
|
| nsv437477 | Chr.6:2934407 - 2948731 on Build GRCh38 | Loss |
|
| nsv600832 | Chr.6:2945097 - 2970334 on Build GRCh38 | Loss |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
| nsv1018369 | Chr.6:2944928 - 2961123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map