Assay Details
Target Gene Details
Entrez Gene ID: | 54821 |
Gene Name: | ERCC excision repair 6 like, spindle assembly checkpoint helicase |
Gene Aliases: |
PICH, RAD26L |
Location: |
Chr.X:72204657-72239008 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERCC6L | NM_017669.2 | 2 | 834 | NP_060139.2 |
| AK056494.1 | 3 | 971 | ||
| AK074719.1 | 1 | 308 | BAC11160.1 | |
| BC111486.1 | 2 | 757 | AAI11487.1 | |
| BM454829.1 | 2 | 757 | ||
| EU069463.1 | 2 | 737 |
Target Gene Details
Entrez Gene ID: | 5303 |
Gene Name: | peptidylprolyl cis/trans isomerase, NIMA-interacting 4 |
Gene Aliases: |
EPVH, PAR14, PAR17 |
Location: |
Chr.X:72181676-72263964 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIN4 | NM_001170747.1 | NP_001164218.1 | ||
| AK127605.1 |
More Information
Set Membership: |
|

Genomic Map