Assay Details
Target Gene Details
Entrez Gene ID: | 201181 |
Gene Name: | zinc finger protein 385C |
Gene Aliases: |
- |
Location: |
Chr.17:42025576-42065668 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF385C | NM_001242704.1 | 8 | 1299 | NP_001229633.1 |
| XM_006721751.3 | 8 | 1407 | XP_006721814.1 | |
| XM_011524479.2 | 9 | 2103 | XP_011522781.1 | |
| XM_011524482.2 | 9 | 2100 | XP_011522784.1 | |
| XM_011524483.2 | 9 | 2089 | XP_011522785.1 | |
| XM_017024314.1 | 10 | 2232 | XP_016879803.1 | |
| XM_017024315.1 | 10 | 2067 | XP_016879804.1 | |
| XM_017024316.1 | 10 | 2229 | XP_016879805.1 | |
| XM_017024317.1 | 10 | 2218 | XP_016879806.1 | |
| XM_017024318.1 | 10 | 2216 | XP_016879807.1 | |
| XM_017024319.1 | 9 | 2086 | XP_016879808.1 | |
| XM_017024320.1 | 9 | 1494 | XP_016879809.1 | |
| BC080613.1 | 4 | 692 | AAH80613.1 | |
| CN264203.1 | ||||
| H06009.1 | 1 | 71 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78965599] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map