Assay Details
Target Gene Details
Entrez Gene ID: | 9510 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 1 |
Gene Aliases: |
C3-C5, METH1 |
Location: |
Chr.21:26836287-26845409 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS1 | NM_006988.4 | 2 | 1477 | NP_008919.3 |
| AB037767.1 | 2 | 1137 | ||
| AF060152.1 | 2 | 1208 | AAD48080.1 | |
| AF170084.1 | 2 | 1264 | AAF15317.1 | |
| AF207664.1 | 2 | 1464 | AAF23772.1 | |
| AK129873.1 | 2 | 328 | BAC85245.1 | |
| AK314475.1 | 2 | 1477 | ||
| BC036515.2 | 2 | 1477 | AAH36515.1 | |
| BX647388.1 | 2 | 1476 | CAI46043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470897 | Chr.21:26823726 - 26843747 on Build GRCh38 | Loss |
|
| nsv834068 | Chr.21:26807351 - 26988489 on Build GRCh38 | Loss |
|
| nsv1066573 | Chr.21:26724037 - 27002035 on Build GRCh38 | Gain |
|
| nsv3482 | Chr.21:26818978 - 26849656 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map