Assay Details
Target Gene Details
Entrez Gene ID: | 4600 |
Gene Name: | MX dynamin like GTPase 2 |
Gene Aliases: |
MXB |
Location: |
Chr.21:41361986-41408943 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MX2 | NM_002463.1 | NP_002454.1 | ||
| XM_005260983.4 | XP_005261040.1 | |||
| XM_005260984.1 | XP_005261041.1 | |||
| XM_011529571.1 | XP_011527873.1 | |||
| XM_011529572.1 | XP_011527874.1 | |||
| XM_011529573.1 | XP_011527875.1 | |||
| AK298780.1 | ||||
| AK308060.1 | ||||
| BC035293.1 | AAH35293.1 | |||
| M30818.1 | AAA36338.1 | |||
| M33883.1 | AAA36459.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528279 | Chr.21:41357910 - 41380411 on Build GRCh38 | Gain |
|
| nsv834102 | Chr.21:41286267 - 41494006 on Build GRCh38 | Loss |
|
| dgv7834n54 | Chr.21:41369022 - 41395967 on Build GRCh38 | Loss |
|
| nsv1057981 | Chr.21:41290295 - 41422831 on Build GRCh38 | Loss |
|
| esv3647060 | Chr.21:41349724 - 41420569 on Build GRCh38 | Gain |
|
| esv3647061 | Chr.21:41354348 - 41391162 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map