Assay Details
Target Gene Details
Entrez Gene ID: | 100131626 |
Gene Name: | uncharacterized LOC100131626 |
Gene Aliases: |
- |
Location: |
Chr.11:118381220-118418815 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100131626 | NR_046369.1 | |||
| NR_046370.1 |
Target Gene Details
Entrez Gene ID: | 9354 |
Gene Name: | ubiquitination factor E4A |
Gene Aliases: |
E4, UBOX2, UFD2 |
Location: |
Chr.11:118359581-118399211 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE4A | NM_001204077.1 | 20 | 5946 | NP_001191006.1 |
| NM_004788.3 | 20 | 5967 | NP_004779.2 | |
| AW021567.1 | ||||
| D50916.1 | 20 | 5904 |
More Information
Set Membership: |
|

Genomic Map