Assay Details
Target Gene Details
Entrez Gene ID: | 23389 |
Gene Name: | mediator complex subunit 13 like |
Gene Aliases: |
MRFACD, PROSIT240, THRAP2, TRAP240L |
Location: |
Chr.12:115958576-116277219 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED13L | NM_015335.4 | 26 | 5846 | NP_056150.1 |
| XM_011538080.2 | 26 | 5915 | XP_011536382.1 | |
| XM_011538081.2 | 26 | 5912 | XP_011536383.1 | |
| XM_011538082.2 | 26 | 6085 | XP_011536384.1 | |
| XM_017019090.1 | 26 | 5876 | XP_016874579.1 | |
| AB028948.2 | 20 | 4913 | BAA82977.2 | |
| AF515599.1 | 26 | 5846 | AAQ08182.1 | |
| AK023837.1 | 10 | 2563 | BAB14697.1 | |
| AL133033.1 | 20 | 4886 | CAB61363.1 | |
| AL137644.1 | 3 | 389 | CAB70855.1 | |
| AY338463.1 | 26 | 5998 | AAR08418.1 | |
| BC130422.1 | 26 | 5842 | ||
| CR749332.1 | 24 | 5370 | CAH18186.1 |
More Information
Set Membership: |
|

Genomic Map