Assay Details
Target Gene Details
Entrez Gene ID: | 5308 |
Gene Name: | paired like homeodomain 2 |
Gene Aliases: |
ARP1, Brx1, IDG2, IGDS, IGDS2, IHG2, IRID2, Otlx2, PTX2, RGS, RIEG, RIEG1, RS |
Location: |
Chr.4:110617423-110642123 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITX2 | NM_000325.5 | 1 | 437 | NP_000316.2 |
| NM_001204397.1 | NP_001191326.1 | |||
| NM_001204398.1 | NP_001191327.1 | |||
| NM_001204399.1 | NP_001191328.1 | |||
| NM_153426.2 | NP_700475.1 | |||
| NM_153427.2 | NP_700476.1 | |||
| XM_011532027.2 | XP_011530329.1 | |||
| AF048720.1 | AAC39716.1 | |||
| AF048721.1 | AAC39717.1 | |||
| AF048722.1 | 1 | 393 | AAC39718.1 | |
| AK127829.1 | ||||
| AK291591.1 | ||||
| AK313987.1 | 1 | 91 | ||
| BC013998.2 | 1 | 437 | AAH13998.1 | |
| BC106010.1 | AAI06011.1 | |||
| BP372081.1 | ||||
| U69961.1 | AAC16257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950378 | Chr.4:110618245 - 110623144 on Build GRCh38 | Deletion |
|
| nsv830036 | Chr.4:110485346 - 110674573 on Build GRCh38 | Gain |
|
| esv2728138 | Chr.4:110565922 - 110650718 on Build GRCh38 | Deletion |
|
| nsv4464 | Chr.4:110592303 - 110625964 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map