Assay Details
Target Gene Details
Entrez Gene ID: | 25839 |
Gene Name: | component of oligomeric golgi complex 4 |
Gene Aliases: |
CDG2J, COD1 |
Location: |
Chr.16:70480569-70523554 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG4 | NM_001195139.1 | NP_001182068.1 | ||
| NM_015386.2 | NP_056201.2 | |||
| XM_011522981.2 | XP_011521283.1 | |||
| AB088369.1 | BAC05682.1 | |||
| AK022874.1 | BAB14286.1 | |||
| AK026435.1 | BAB15483.1 | |||
| AK096557.1 | BAC04816.1 | |||
| AK293235.1 | ||||
| AK297557.1 | ||||
| AL050101.1 | CAB43272.1 | |||
| BC006306.2 | AAH06306.2 | |||
| BC013347.2 | AAH13347.2 | |||
| BC027726.2 | AAH27726.1 | |||
| BC072438.1 | AAH72438.1 | |||
| BC128144.1 | ||||
| BC128145.1 | ||||
| BX648010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833272 | Chr.16:70335866 - 70535820 on Build GRCh38 | Loss |
|
| esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map