Assay Details
Target Gene Details
Entrez Gene ID: | 2674 |
Gene Name: | GDNF family receptor alpha 1 |
Gene Aliases: |
GDNFR, GDNFRA, GFR-ALPHA-1, RET1L, RETL1, TRNR1 |
Location: |
Chr.10:116056925-116273645 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRA1 | NM_001145453.2 | 10 | 2024 | NP_001138925.1 |
| NM_005264.5 | 11 | 2039 | NP_005255.1 | |
| NM_145793.3 | 9 | 1573 | NP_665736.1 | |
| XM_005269696.1 | 11 | 1649 | XP_005269753.1 | |
| XM_011539634.1 | 11 | 1614 | XP_011537936.1 | |
| AF038421.1 | 11 | 1866 | AAC39692.1 | |
| AF042080.1 | 10 | 1420 | AAB97371.1 | |
| AK292886.1 | 10 | 1585 | ||
| AK302914.1 | 7 | 1039 | ||
| BC014962.1 | 9 | 1548 | AAH14962.1 | |
| BU935138.1 | 3 | 142 | ||
| U95847.1 | 9 | 1539 | AAB71811.1 | |
| U97144.1 | 9 | 1556 | AAC51646.1 |
More Information
Set Membership: |
|

Genomic Map