Assay Details
Target Gene Details
Entrez Gene ID: | 341640 |
Gene Name: | FRAS1 related extracellular matrix protein 2 |
Gene Aliases: |
- |
Location: |
Chr.13:38687036-38887131 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FREM2 | NM_207361.5 | 1 | 1619 | NP_997244.4 |
| XM_017020554.1 | 1 | 1614 | XP_016876043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832585 | Chr.13:38659170 - 38850238 on Build GRCh38 | Gain |
|
| nsv832584 | Chr.13:38500128 - 38710587 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map