Assay Details
Target Gene Details
Entrez Gene ID: | 6403 |
Gene Name: | selectin P |
Gene Aliases: |
CD62, CD62P, GMP140, GRMP, LECAM3, PADGEM, PSEL |
Location: |
Chr.1:169588849-169630139 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SELP | NM_003005.3 | 17 | 2799 | NP_002996.2 |
| XM_005245435.1 | 17 | 2900 | XP_005245492.1 | |
| XM_005245438.1 | 16 | 2780 | XP_005245495.1 | |
| XM_005245439.1 | 16 | 2676 | XP_005245496.1 | |
| XM_005245440.1 | 16 | 2714 | XP_005245497.1 | |
| BC028067.2 | 15 | 2715 | AAH28067.2 | |
| BC068533.1 | 17 | 2782 | AAH68533.1 | |
| M25322.1 | 17 | 2772 | AAA35911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548187 | Chr.1:169561599 - 169589797 on Build GRCh38 | Loss |
|
| esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map