Assay Details
Target Gene Details
Entrez Gene ID: | 57534 |
Gene Name: | mindbomb E3 ubiquitin protein ligase 1 |
Gene Aliases: |
DIP-1, DIP1, LVNC7, MIB, ZZANK2, ZZZ6 |
Location: |
Chr.18:21740793-21870957 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIB1 | NM_020774.3 | 1 | 404 | NP_065825.1 |
| XM_017025874.1 | 1 | 940 | XP_016881363.1 | |
| XM_017025875.1 | 1 | 940 | XP_016881364.1 | |
| AY147849.1 | 1 | 421 | AAN18023.1 | |
| AY149908.1 | 1 | 149 | AAN75493.1 | |
| BC110581.1 | 1 | 149 | AAI10582.1 | |
| BC110582.1 | 1 | 149 | AAI10583.1 |
More Information
Set Membership: |
|

Genomic Map