Assay Details
Target Gene Details
Entrez Gene ID: | 388611 |
Gene Name: | CD164 molecule like 2 |
Gene Aliases: |
EAPG6122, UNQ6122 |
Location: |
Chr.1:27379100-27383333 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD164L2 | NM_207397.3 | 5 | 1371 | NP_997280.2 |
| XM_005245868.3 | 6 | 812 | XP_005245925.2 | |
| XM_011541441.1 | XP_011539743.1 | |||
| AY358761.1 | 5 | 1341 | AAQ89121.1 | |
| BC137466.1 | ||||
| BM145906.1 | 1 | 282 | ||
| BM145910.1 | 1 | 282 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526051 | Chr.1:27284255 - 27445496 on Build GRCh38 | Gain |
|
| esv1006916 | Chr.1:27366576 - 27380519 on Build GRCh38 | Deletion |
|
| nsv516766 | Chr.1:27335744 - 27405147 on Build GRCh38 | Loss |
|

Genomic Map