Assay Details
Target Gene Details
Entrez Gene ID: | 89978 |
Gene Name: | diphthamine biosynthesis 6 |
Gene Aliases: |
ATPBD4 |
Location: |
Chr.15:35217356-35546222 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH6 | NM_001141972.1 | NP_001135444.1 | ||
| NM_080650.3 | NP_542381.1 | |||
| XM_011522160.1 | XP_011520462.1 | |||
| XM_017022707.1 | XP_016878196.1 | |||
| XM_017022708.1 | XP_016878197.1 | |||
| XM_017022709.1 | XP_016878198.1 | |||
| AK094007.1 | BAC04266.1 | |||
| AK125017.1 | ||||
| AL833427.1 | ||||
| BC066652.1 | AAH66652.1 | |||
| DB250468.1 |
Target Gene Details
Entrez Gene ID: | 100507466 |
Gene Name: | DPH6 antisense RNA 1 (head to head) |
Gene Aliases: |
ATPBD4-AS1 |
Location: |
Chr.15:35546195-35859001 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH6-AS1 | NR_038251.1 |
More Information
Set Membership: |
|

Genomic Map