Assay Details
Target Gene Details
Entrez Gene ID: | 57680 |
Gene Name: | chromodomain helicase DNA binding protein 8 |
Gene Aliases: |
AUTS18, HELSNF1 |
Location: |
Chr.14:21385194-21437298 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHD8 | NM_001170629.1 | 33 | 6662 | NP_001164100.1 |
| NM_020920.3 | 34 | 5907 | NP_065971.2 | |
| AB046784.3 | 33 | 6150 | BAB13390.3 | |
| AK056639.1 | 14 | 2529 | ||
| AK131077.1 | 19 | 3486 | BAC85127.1 | |
| BC011695.2 | 2 | 208 | AAH11695.2 | |
| BC025964.1 | 2 | 208 | AAH25964.1 | |
| BC063693.1 | 2 | 226 | AAH63693.1 | |
| BC073903.1 | 3 | 332 | AAH73903.1 | |
| BC098452.1 | 8 | 1578 | AAH98452.1 | |
| CR749315.1 | 34 | 5852 | CAH18170.1 |
More Information
Set Membership: |
|

Genomic Map