Assay Details
Target Gene Details
Entrez Gene ID: | 1823 |
Gene Name: | desmocollin 1 |
Gene Aliases: |
CDHF1, DG2/DG3 |
Location: |
Chr.18:31129236-31162856 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSC1 | NM_004948.3 | 17 | 3486 | NP_004939.1 |
| NM_024421.2 | 16 | 3440 | NP_077739.1 | |
| X72925.1 | 17 | 3486 | CAA51429.1 | |
| Z34522.1 | 17 | 3414 | CAA84278.1 |
Target Gene Details
Entrez Gene ID: | 101927698 |
Gene Name: | DSC1/DSC2 antisense RNA |
Gene Aliases: |
- |
Location: |
Chr.18:31101588-31162789 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSCAS | NR_110785.1 | |||
| DA156712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519734 | Chr.18:31127452 - 31141571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map