Assay Details
Target Gene Details
Entrez Gene ID: | 219287 |
Gene Name: | APC membrane recruitment protein 2 |
Gene Aliases: |
FAM123A |
Location: |
Chr.13:25161679-25172456 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMER2 | NM_152704.3 | 1 | 3244 | NP_689917.2 |
| NM_199138.2 | 3 | 2578 | NP_954589.1 | |
| XM_005266279.3 | 2 | 3224 | XP_005266336.1 | |
| XM_005266280.3 | 2 | 3134 | XP_005266337.1 | |
| XM_017020415.1 | 2 | 2898 | XP_016875904.1 | |
| AK098796.1 | 2 | 1519 | BAC05414.1 | |
| AL832627.1 | 1 | 1297 | ||
| BM680074.1 | 1 | 523 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582237 | Chr.13:25162386 - 25434284 on Build GRCh38 | Gain |
|
| esv2747161 | Chr.13:25031743 - 26209531 on Build GRCh38 | Deletion |
|
| nsv826594 | Chr.13:25167847 - 25169194 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map