Assay Details
Target Gene Details
Entrez Gene ID: | 25897 |
Gene Name: | ring finger protein 19A, RBR E3 ubiquitin protein ligase |
Gene Aliases: |
RNF19 |
Location: |
Chr.8:100257059-100336218 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF19A | NM_001280539.1 | NP_001267468.1 | ||
| NM_015435.4 | NP_056250.3 | |||
| NM_183419.3 | NP_904355.1 | |||
| XM_005250853.3 | XP_005250910.1 | |||
| XM_011516956.2 | XP_011515258.1 | |||
| XM_017013302.1 | XP_016868791.1 | |||
| XM_017013303.1 | XP_016868792.1 | |||
| XM_017013304.1 | XP_016868793.1 | |||
| XM_017013305.1 | XP_016868794.1 | |||
| AB029316.1 | BAB39353.1 | |||
| BC093938.1 | AAH93938.1 | |||
| BC093940.2 | AAH93940.1 |
More Information
Set Membership: |
|

Genomic Map