Assay Details
Target Gene Details
Entrez Gene ID: | 51802 |
Gene Name: | acid sensing ion channel subunit family member 5 |
Gene Aliases: |
ACCN5, HINAC, INAC |
Location: |
Chr.4:155829699-155867303 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASIC5 | NM_017419.2 | NP_059115.1 | ||
| XM_017008291.1 | XP_016863780.1 | |||
| AJ252011.1 | CAB85607.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029122 | Chr.4:155773161 - 156031641 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map