Assay Details
Target Gene Details
Entrez Gene ID: | 54865 |
Gene Name: | G-patch domain containing 4 |
Gene Aliases: |
GPATC4 |
Location: |
Chr.1:156594308-156601496 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPATCH4 | NM_015590.3 | 8 | 1458 | NP_056405.2 |
| NM_182679.2 | 8 | 1532 | NP_872620.1 | |
| XM_005245287.3 | 7 | 1454 | XP_005245344.1 | |
| AK293026.1 | 8 | 1526 | ||
| BM997996.1 | 1 | 458 |
Target Gene Details
Entrez Gene ID: | 128240 |
Gene Name: | NAD(P)HX epimerase |
Gene Aliases: |
AIBP, APOA1BP, YJEFN1 |
Location: |
Chr.1:156591762-156599818 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAXE | XM_017000319.1 | XP_016855808.1 | ||
| AK294835.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523181 | Chr.1:156580462 - 156706761 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77181873] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map