Assay Details
Target Gene Details
Entrez Gene ID: | 64844 |
Gene Name: | membrane associated ring-CH-type finger 7 |
Gene Aliases: |
AXO, AXOT, MARCH-VII, RNF177 |
Location: |
Chr.2:159712457-159768583 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARCH7 | NM_001282805.1 | 4 | 335 | NP_001269734.1 |
| NM_001282806.1 | 2 | 207 | NP_001269735.1 | |
| NM_022826.3 | 2 | 207 | NP_073737.1 | |
| XM_005246773.2 | 2 | 230 | XP_005246830.1 | |
| XM_005246774.1 | 2 | 230 | XP_005246831.1 | |
| XM_017004718.1 | 2 | 207 | XP_016860207.1 | |
| XM_017004719.1 | 2 | 233 | XP_016860208.1 | |
| XM_017004720.1 | 2 | 195 | XP_016860209.1 | |
| XM_017004721.1 | 2 | 225 | XP_016860210.1 | |
| AK022973.1 | 2 | 165 | BAB14340.1 | |
| AK222880.1 | 2 | 157 | BAD96600.1 | |
| AK292836.1 | 4 | 303 | ||
| AK299051.1 | 2 | 175 | ||
| BC065014.1 | 2 | 131 | AAH65014.1 | |
| DA825046.1 | 2 | 211 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010774 | Chr.2:159012247 - 160175703 on Build GRCh38 | Gain |
|
| nsv470497 | Chr.2:159694632 - 159836250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map