Assay Details
Target Gene Details
Entrez Gene ID: | 23387 |
Gene Name: | SIK family kinase 3 |
Gene Aliases: |
L19, QSK, SIK-3 |
Location: |
Chr.11:116843402-117098453 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIK3 | NM_001281748.1 | 21 | 3305 | NP_001268677.1 |
| NM_001281749.1 | 20 | 3181 | NP_001268678.1 | |
| NM_025164.4 | 20 | 3361 | NP_079440.3 | |
| XM_005271481.2 | 21 | 3543 | XP_005271538.1 | |
| XM_005271482.4 | 20 | 3399 | XP_005271539.2 | |
| XM_005271484.3 | 20 | 3219 | XP_005271541.2 | |
| XM_005271485.3 | 8 | 1881 | XP_005271542.1 | |
| XM_011542721.2 | 21 | 3505 | XP_011541023.1 | |
| XM_011542722.1 | 21 | 3363 | XP_011541024.1 | |
| XM_011542723.2 | 21 | 3386 | XP_011541025.1 | |
| XM_011542724.2 | 20 | 3240 | XP_011541026.1 | |
| XM_011542725.2 | 19 | 3123 | XP_011541027.1 | |
| XM_017017424.1 | 19 | 3075 | XP_016872913.1 | |
| XM_017017425.1 | 21 | 3581 | XP_016872914.1 | |
| XM_017017426.1 | 19 | 3455 | XP_016872915.1 | |
| XM_017017427.1 | 16 | 2858 | XP_016872916.1 | |
| AB209327.1 | 13 | 2371 | BAD92564.1 | |
| AK022302.1 | 2 | 916 | BAB14006.1 | |
| AK122838.1 | 6 | 2623 | ||
| AY598338.1 | 2 | 913 | AAT06749.1 | |
| BC035583.2 | 3 | 1191 | AAH35583.3 | |
| BC063887.1 | 6 | 1374 | AAH63887.1 | |
| BC086858.1 | 2 | 916 | AAH86858.1 | |
| BC128510.1 | 20 | 3043 | ||
| BC128511.1 | 21 | 3161 |
More Information
Set Membership: |
|

Genomic Map