Assay Details
Target Gene Details
Entrez Gene ID: | 131890 |
Gene Name: | G protein-coupled receptor kinase 7 |
Gene Aliases: |
GPRK7 |
Location: |
Chr.3:141763127-141817373 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK7 | NM_139209.2 | 2 | 814 | NP_631948.1 |
| XM_017005717.1 | 4 | 3556 | XP_016861206.1 | |
| AF282269.1 | 2 | 814 | AAL33880.1 | |
| AF439409.1 | 2 | 867 | AAL48216.1 |
More Information
Set Membership: |
|

Genomic Map