Assay Details
Target Gene Details
Entrez Gene ID: | 3476 |
Gene Name: | immunoglobulin (CD79A) binding protein 1 |
Gene Aliases: |
ALPHA-4, IBP1 |
Location: |
Chr.X:70133449-70166324 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGBP1 | NM_001551.2 | NP_001542.1 | ||
| XM_017029489.1 | 1 | 66 | XP_016884978.1 | |
| XM_017029490.1 | 1 | 66 | XP_016884979.1 | |
| AK054596.1 | 1 | 66 | ||
| AK315660.1 | ||||
| BC004137.2 | AAH04137.1 |
More Information
Set Membership: |
|

Genomic Map