Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C12orf60 | NM_175874.3 | NP_787070.2 | ||
| XM_005253322.4 | XP_005253379.1 | |||
| XM_017018871.1 | XP_016874360.1 | |||
| XM_017018872.1 | XP_016874361.1 | |||
| XM_017018873.1 | XP_016874362.1 | |||
| XM_017018874.1 | XP_016874363.1 | |||
| AK056213.1 | ||||
| AK289942.1 | ||||
| BC015546.1 | AAH15546.1 | |||
| BC029443.1 | AAH29443.1 | |||
| BC038836.1 | AAH38836.1 |
Target Gene Details
Entrez Gene ID: | 440087 |
Gene Name: | single-pass membrane protein with coiled-coil domains 3 |
Gene Aliases: |
C12orf69 |
Location: |
Chr.12:14804650-14816761 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMCO3 | NM_001013698.2 | 2 | 1739 | NP_001013720.2 |
| XM_017019312.1 | 3 | 4404 | XP_016874801.1 | |
| AK092637.1 | 2 | 1739 | BAC03930.1 | |
| BU735185.1 | 1 | 203 |
More Information
Set Membership: |
|

Genomic Map