Assay Details
Target Gene Details
Entrez Gene ID: | 8642 |
Gene Name: | dachsous cadherin-related 1 |
Gene Aliases: |
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1 |
Location: |
Chr.11:6621324-6655849 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCHS1 | NM_003737.3 | 21 | 8673 | NP_003728.1 |
| AB053446.1 | 21 | 8676 | ||
| AL831828.1 | 8 | 2495 | CAD38541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951304 | Chr.11:6620670 - 6634169 on Build GRCh38 | Deletion |
|
| nsv553373 | Chr.11:6601484 - 6647097 on Build GRCh38 | Loss |
|
| nsv1035138 | Chr.11:6607812 - 6980555 on Build GRCh38 | Gain |
|
| nsv553374 | Chr.11:6616273 - 6664327 on Build GRCh38 | Loss |
|
| nsv825755 | Chr.11:6591998 - 6656613 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map