Assay Details
Target Gene Details
Entrez Gene ID: | 23107 |
Gene Name: | mitochondrial ribosomal protein S27 |
Gene Aliases: |
MRP-S27, S27mt |
Location: |
Chr.5:72219409-72320257 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS27 | NM_001286748.1 | 12 | 2672 | NP_001273677.1 |
| NM_001286751.1 | 11 | 2598 | NP_001273680.1 | |
| NM_015084.2 | 11 | 2630 | NP_055899.2 | |
| AK025417.1 | ||||
| AK027001.1 | ||||
| AK128683.1 | ||||
| BC003632.2 | AAH03632.1 | |||
| BC030521.2 | AAH30521.1 | |||
| BC064902.1 | AAH64902.1 | |||
| BM995930.1 | 1 | 168 | ||
| D87453.1 |
More Information
Set Membership: |
|

Genomic Map