Assay Details
Target Gene Details
Entrez Gene ID: | 84167 |
Gene Name: | chromosome 19 open reading frame 44 |
Gene Aliases: |
- |
Location: |
Chr.19:16496311-16521369 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C19orf44 | NM_032207.3 | NP_115583.1 | ||
| XM_006722920.3 | XP_006722983.1 | |||
| AK025395.1 | BAB15125.1 | |||
| AK297781.1 | ||||
| BC027869.1 | AAH27869.1 |
Target Gene Details
Entrez Gene ID: | 10523 |
Gene Name: | calcium homeostasis endoplasmic reticulum protein |
Gene Aliases: |
DAN16, SCAF6, SRA1 |
Location: |
Chr.19:16517889-16542452 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHERP | NM_006387.5 | 17 | 2915 | NP_006378.3 |
| AB209730.1 | 8 | 1292 | BAD92967.1 | |
| AF536542.1 | 17 | 2882 | AAN77183.1 | |
| AK296883.1 | 11 | 1626 | ||
| AK298408.1 | 9 | 1643 | ||
| BC021294.2 | 17 | 2900 | AAH21294.1 | |
| U94836.2 | 16 | 2817 | AAB53327.1 |
More Information
Set Membership: |
|

Genomic Map