Assay Details
Target Gene Details
Entrez Gene ID: | 55773 |
Gene Name: | TBC1 domain family member 23 |
Gene Aliases: |
NS4ATP1 |
Location: |
Chr.3:100260817-100325252 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D23 | NM_001199198.2 | 1 | 206 | NP_001186127.1 |
| NM_018309.4 | 1 | 206 | NP_060779.2 | |
| XM_017006840.1 | XP_016862329.1 | |||
| XM_017006841.1 | XP_016862330.1 | |||
| AB449910.1 | ||||
| AK001908.1 | BAA91973.1 | |||
| BC020955.1 | AAH20955.1 | |||
| BX648032.1 | ||||
| DA801258.1 | 1 | 206 | ||
| DB458401.1 | 1 | 181 |
More Information
Set Membership: |
|

Genomic Map