Assay Details
Target Gene Details
Entrez Gene ID: | 372 |
Gene Name: | archain 1 |
Gene Aliases: |
COPD |
Location: |
Chr.11:118572363-118603033 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARCN1 | NM_001142281.1 | NP_001135753.1 | ||
| NM_001655.4 | 2 | 215 | NP_001646.2 | |
| XM_005271542.3 | 2 | 239 | XP_005271599.1 | |
| AK304019.1 | ||||
| AK312298.1 | 2 | 193 | ||
| BC064936.1 | 2 | 178 | AAH64936.1 | |
| BC093636.1 | 2 | 142 | AAH93636.1 | |
| BC093638.1 | 2 | 139 | AAH93638.1 | |
| BI560739.1 | 2 | 211 | ||
| BX640860.1 | 3 | 343 | CAE45922.1 | |
| DB216489.1 | 2 | 215 | ||
| X81197.1 | 2 | 171 | CAA57071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832283 | Chr.11:118566790 - 118786484 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map